Anti-ALDH3A1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106191
89319-564EA 394.91 USD
89319-564
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GSC (goosecoid homeobox)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ALDH3A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR