Anti-MEP1A Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106151
89319-558EA 394.91 USD
89319-558
Anti-MEP1A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Meprin A alpha (meprin A, alpha (PABA peptide hydrolase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MEP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR