Anti-BHMT Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106029
89319-530EA 394.79 USD
89319-530
Anti-BHMT Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ADPGK (ADP-dependent glucokinase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BHMT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR