Anti-BRAF Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX106004
89319-522EA 427.1 USD
89319-522
Anti-BRAF Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to UTP6 (UTP6, small subunit (SSU) processome component, homolog (yeast))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BRAF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR