Anti-BSG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105933
89319-504EA 582.76 USD
89319-504
Anti-BSG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PSKH2 (protein serine kinase H2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BSG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR