Anti-BSG Goat Polyclonal Antibody

Supplier: GeneTex
GTX10590
89285-976EA 608.06 USD
89285-976
Anti-BSG Goat Polyclonal Antibody
Antibodies
Goat polyclonal to RANTES

Recommended Starting Dilutions:

For WB: Use at a concentration of 0.1 ?g/ml.

For IHC-Fr: Use at a concentration of 5-15 ?g/ml.

For Neutralization: The ND50 is 0.1-0.4 µg/mL in the presence of 0.01 µg/mL of recombinant human RANTES, using human CCR5 transfected BAF3 cells.

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: BSG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR