Anti-BTK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105893
89319-498EA 482.37 USD
89319-498
Anti-BTK Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MEK6 (mitogen-activated protein kinase kinase 6)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BTK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR