Anti-BUB1B Rabbit Polyclonal Antibody

GTX105815
89309-278EA 176.03 USD
89309-278
Anti-BUB1B Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Carbonic anhydrase 14 (carbonic anhydrase XIV)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: BUB1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR