Anti-BUB1B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105811
89335-402EA 582.76 USD
89335-402
Anti-BUB1B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD3 delta (CD3d molecule, delta (CD3-TCR complex))

Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BUB1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR