Anti-C16ORF72 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105780
89319-478EA 394.79 USD
89319-478
Anti-C16ORF72 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ASC1 (thyroid hormone receptor interactor 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C16ORF72
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR