Anti-C9orf41 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105742
89319-470EA 394.91 USD
89319-470
Anti-C9orf41 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PVRL2(CD112) (poliovirus receptor-related 2 (herpesvirus entry mediator B))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C9orf41
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR