Anti-C9orf41 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105736
89309-250EA 394.91 USD
89309-250
Anti-C9orf41 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FUCA2 (fucosidase, alpha-L- 2, plasma)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: C9orf41
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR