Anti-CD4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105666
89319-452EA 641.7 USD
89319-452
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to KCNMB1 (potassium large conductance calcium-activated channel, subfamily M, beta member 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR