Anti-CDX2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105625
89319-446EA 376.43 USD
89319-446
Anti-CDX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GEF-H1 (rho/rac guanine nucleotide exchange factor (GEF) 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDX2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR