Anti-CLPP Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105584
89319-434EA 394.91 USD
89319-434
Anti-CLPP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Casein Kinase 1 alpha 1L (casein kinase 1, alpha 1-like)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CLPP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR