Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COX6A1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105550
89319-428EA 358.82 USD InStock
89319-428
Anti-COX6A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZNF198 (zinc finger, MYM-type 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:50-1:500, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: COX6A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty