Anti-CRYAB Rabbit Polyclonal Antibody

GTX105490
89309-158EA 176.03 USD
89309-158
Anti-CRYAB Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to ZNF175 (zinc finger protein 175)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR