Anti-CRYAB Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105470
89319-418EA 394.79 USD
89319-418
Anti-CRYAB Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZNF449 (zinc finger protein 449)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR