Anti-CTNNB1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105433
89335-394EA 597.31 USD
89335-394
Anti-CTNNB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MEF2C (myocyte enhancer factor 2C)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR