Anti-CYB5R3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105412
89330-420EA 404.66 USD
89330-420
Anti-CYB5R3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SIGIRR (single immunoglobulin and toll-interleukin 1 receptor (TIR) domain)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR