Anti-CYP1A2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX105380
89319-404EA 427.1 USD
89319-404
Anti-CYP1A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NKRF (NFKB repressing factor)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYP1A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR