Anti-CYP2E1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105373
89319-402EA 394.91 USD
89319-402
Anti-CYP2E1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ASIC1 (amiloride-sensitive cation channel 2, neuronal)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYP2E1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR