Anti-DAPK2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105344
89319-398EA 582.76 USD
89319-398
Anti-DAPK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PWP2 (PWP2 periodic tryptophan protein homolog (yeast))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DAPK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR