Anti-DCXR Rabbit Polyclonal Antibody

GTX105325
89309-134EA 176.03 USD
89309-134
Anti-DCXR Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to RP5-1022P6.2

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: DCXR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR