Anti-DCXR Rabbit Polyclonal Antibody

GTX105320
89309-132EA 176.03 USD
89309-132
Anti-DCXR Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to ENC1 (ectodermal-neural cortex (with BTB-like domain))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: DCXR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR