Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNAJ Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105283
89309-122EA 167.65 USD InStock
89309-122
Anti-DNAJ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to eIF3 eta (eukaryotic translation initiation factor 3, subunit B)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: DNAJ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty