Anti-DUPD1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105239
89319-386EA 582.76 USD
89319-386
Anti-DUPD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Leukotriene A4 hydrolase (leukotriene A4 hydrolase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DUPD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR