Anti-ELAVL1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105174
89319-368EA 582.76 USD
89319-368
Anti-ELAVL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TAF7 (TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ELAVL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR