Anti-EPCAM Rabbit Polyclonal Antibody

Supplier: Genetex

GTX105162
89309-078EA 793.5 USD
89309-078
Anti-EPCAM Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CNGA3 (cyclic nucleotide gated channel alpha 3)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: EPCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR