Anti-ERCC1 Goat Polyclonal Antibody

Supplier: GeneTex

GTX10507
89317-202EA 427.58 USD
89317-202
Anti-ERCC1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to TNFSF5

Recommended Starting Dilutions:

For WB: Use at a concentration of 1 ?g/mL.

For FACS: Use at an assay dependent dilution.

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ERCC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR