Anti-FAHD2A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX105037
89319-338EA 582.76 USD
89319-338
Anti-FAHD2A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to METTL3 (methyltransferase like 3)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: METTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR