Anti-FCGR2A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104949
89319-314EA 582.76 USD
89319-314
Anti-FCGR2A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PRPF39 (PRP39 pre-mRNA processing factor 39 homolog (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRPF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR