Anti-FERMT2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104918
89335-382EA 582.76 USD
89335-382
Anti-FERMT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ELA3A (chymotrypsin-like elastase family, member 3A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CELA3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR