Anti-FLT3LG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104888
89319-292EA 582.76 USD
89319-292
Anti-FLT3LG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CLIC4 (chloride intracellular channel 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CLIC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR