Anti-FRK Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104873
89319-286EA 394.91 USD
89319-286
Anti-FRK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to KIF4A (kinesin family member 4A)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KIF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR