Anti-GAD1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104851
89308-986EA 176.03 USD
89308-986
Anti-GAD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NERF (E74-like factor 2 (ets domain transcription factor))

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ELF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR