Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAS7 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104846
89308-980EA 167.65 USD InStock
89308-980
Anti-GAS7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FOXD2 (forkhead box D2)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: FOXD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty