Anti-GATA6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104833
89316-442EA 395.48 USD
89316-442
Anti-GATA6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to KLF1 (Kruppel-like factor 1 (erythroid))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KLF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR