Anti-GBP2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX104810
89319-274EA 427.1 USD
89319-274
Anti-GBP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to DAP Kinase 2 (death-associated protein kinase 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR