Anti-GFAP Rabbit Polyclonal Antibody

GTX104802
89308-934EA 176.03 USD
89308-934
Anti-GFAP Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SENP6 (SUMO1/sentrin specific peptidase 6)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SENP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR