Anti-GSTT2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104736
89308-902EA 394.91 USD
89308-902
Anti-GSTT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to DOT1L (DOT1-like, histone H3 methyltransferase (S. cerevisiae))

Suggested starting dilutions are as follows: WB: Assay dependent. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DOT1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR