Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDHD2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104689
89308-870EA 167.65 USD InStock
89308-870
Anti-HDHD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RPL38 (ribosomal protein L38)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: HDHD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty