Anti-HP Rabbit Polyclonal Antibody

GTX104641
89319-206EA 376.76 USD
89319-206
Anti-HP Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to USP16 (ubiquitin specific peptidase 16)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR