Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-HSPA6 Goat Polyclonal Antibody

Supplier: Genetex

GTX10459
89317-188EA 359.14 USD InStock
89317-188
Anti-HSPA6 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to CD137

ELISA: Use at a concentration of 0.5 - 1.0 μg/ml to detect 0.06 ng/well of human CD137. WB: Use at a concentration of 0.1 - 0.2 μg/ml to detect 0.2 ng/lane and 0.5 ng/lane under non-reducing and reducing conditions, respectively. Predicted molecular weight: 28 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HSPA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty