Anti-HSPA9 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104579
89330-414EA 582.76 USD
89330-414
Anti-HSPA9 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FUBP1 (far upstream element-binding protein 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HSPA9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR