Anti-IDH3A Rabbit Polyclonal Antibody

Supplier: Genetex

GTX10452
89286-762EA 395.36 USD
89286-762
Anti-IDH3A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Caspase 10

WB: Use at a dilution of 1/500. Predicted molecular weight: 59 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IDH3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR