Anti-IFIT3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10451
89286-760EA 455.16 USD
89286-760
Anti-IFIT3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Caspase 8

WB: Use at a dilution of 1/1000 (determined by using a whole cell extract of the human Burkitt lymphoma Raji cell line). Anti-Caspase 8 (GD-13) recognizes human caspase 8a (55 kDa) and caspase 8b (54kDa) by immunoblotting. Staining of caspase 8 in immunoblotting is specifically inhibited with caspase 8 immunizing peptide. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IFIT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR