Anti-IFT57 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10450
89286-758EA 436.11 USD
89286-758
Anti-IFT57 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Caspase 8

WB: Use at a dilution of 1/1000 (determined by using a whole cell extract of the human Burkitt lymphoma Raji cell line). Expected bands at: 55 kDa (human caspase 8a) and 54 kDa (caspase 8b). Possible bands at 30-32 kDa (caspase 8 MACH beta 3/beta 4 isoforms). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IFT57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR