Anti-IFT57 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104491
89308-806EA 394.91 USD
89308-806
Anti-IFT57 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Kallikrein 6 (kallikrein-related peptidase 6)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IFT57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR