Anti-IL1F6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104422
89319-174EA 582.56 USD
89319-174
Anti-IL1F6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PSME1 (proteasome (prosome, macropain) activator subunit 1 (PA28 alpha))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IL1F6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR